Learn More
Aviva Systems Biology Corp KLHL25 Antibody - N-terminal region (ARP34830_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34830_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL25. Alternate names: ENC-2, ENC2, FLJ12587ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-55579