Learn More
Aviva Systems Biology Corp KLHL26 Antibody - middle region (ARP34578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL26. Alternate names: FLJ11078ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-55460