Learn More
Aviva Systems Biology Corp KLHL3 Antibody - N-terminal region (ARP39276_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39276_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL3. Alternate names: PHA2DApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-56967