Learn More
Aviva Systems Biology Corp KLHL31 Antibody - C-terminal region (ARP44458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL31. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human KLHL31. Alternate names: BKLHD6, KLHL, bA345L23.2, KBTBD1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58817