Learn More
Aviva Systems Biology Corp KLHL32 Antibody - middle region (ARP52449_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52449_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL32. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL32. Alternate names: BKLHD5, KIAA1900, MGC51280, MGC87753, RP1-39B17.1, UG0030H05, dJ21F7.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60744