Learn More
Aviva Systems Biology Corp KLHL4 Antibody - N-terminal region (ARP39386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLHL4. Alternate names: DKELCHL, KHL4, KIAA1687ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57013