Learn More
Aviva Systems Biology Corp KLHL7 Antibody - middle region (ARP56234_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56234_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL7. Alternate names: KLHL6, SBBI26ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61775