Learn More
Aviva Systems Biology Corp KLHL9 Antibody - middle region (ARP57295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLHL9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLHL9. Alternate names: FLJ21815, RP11-380P16.6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62121