Learn More
Aviva Systems Biology Corp KLK13 Antibody - middle region (ARP55295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLK13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLK13. Alternate names: DKFZP586J1923, KLK-L4, KLKL4ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53911