Learn More
Aviva Systems Biology Corp KLK2 Antibody - middle region (ARP41842_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41842_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLK2. Alternate names: KLK2A2, MGC12201, hK2, hGK-1ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-58016