Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 354MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: KLK3/1248. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: PC12 cells or normal prostate or prostate carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human KLK3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 33-34kDa.Chromosome: 19q13.33. Gene Symbol: KLK3. Gene ID: 354. Ensemble: ENSG00000142515. Uniprot: NP_001639.
Catalog No. 50-188-4825