Learn More
Aviva Systems Biology Corp KLK3 Antibody - middle region (ARP41879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLK3. It was validated on Western Blot. . Alternate names: APS, KLK2A1, PSA, hK3ApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-284-962