Learn More
Aviva Systems Biology Corp KLK5 Antibody - N-terminal region (ARP45010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLK5. Alternate names: KLK-L2, KLKL2, SCTEApplicationWB. Species reactivity: Human
Catalog No. 50-014-58952