Learn More
Aviva Systems Biology Corp KLK6 Antibody - N-terminal region (ARP33888_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33888_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLK6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLK6. Alternate names: Bssp, Klk7, MGC9355, NEUROSIN, PRSS18, PRSS9, SP59, ZYME, hK6ApplicationIHC, WB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-55188