Learn More
Aviva Systems Biology Corp KLKB1 Antibody - middle region (ARP45678_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45678_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against KLKB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLKB1. Alternate names: KLK3, PPKApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-53450