Learn More
MCR Safety™ Klondike KD7 Black/Gray, Clear UV-AF
Catalog No. 19827922 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-827-922 Anti-fog, Impact-resistant
1 options
Catalog No. 19-827-922 Supplier MCR Safety™ Supplier No. KD710AF
Klondike™ KD7 Series - Black frame, gray and black temples, extremely low profile and lightweight, zero removable parts, clear UV-AF™ anti-fog lens.
  • Available with UV-AF™, MAX6™, or MAX36™ lens coatings
  • Extremely low profile, lightweight (24.8 grams) half-frame design
  • Zero removable components
  • Rugged polycarbonate lenses filter 99.9% of harmful U.V. rays (385nm)
  • Streamlined, permanently bonded TPR temple arms
  • Soft, permanently bonded TPR nose pad with adjustable customized fit
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Clear
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Half Frame
Autoclavable Not Autoclavable