Learn More
MCR Safety™ Klondike OTG Safety Glasses, Black/Gray, Clear MAX6
Catalog No. 19827921 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-827-921 Anti-fog
1 options
Catalog No. 19-827-921 Supplier MCR Safety™ Supplier No. OG210PF
Klondike™ OTG Series - Over-the-Glass safety glasses, clear MAX6™ anti-fog lens, black frame and black temples, 3 position adjustable temple length.
  • 3-position adjustable temple length
  • Available with MAX6™ or MAX36™ lens coatings
  • Oversized frame and lens to fit over prescription glasses
  • Rugged polycarbonate lenses filter 99.9% of harmful U.V. rays (385nm)
  • Soft, permanently bonded TPR browguard to keep out debris
  • Streamlined, permanently bonded TPR temple arms
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Overglasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless
Autoclavable Not Autoclavable