Learn More
Medchemexpress LLC Klotho beta Human 10ug | 10ug
SDP
Catalog No. 5000195341
Encompass_Preferred
Klotho beta protein plays a critical role in the transcriptional repression of cholesterol 7-alpha-hydroxylase (CYP7A1) a key enzyme controlling bile acid synthesis Although its glycosidase activity may be inactive Klotho beta enhances the binding ability of FGFR1 and FGFR4 to FGF21 promoting their interaction Klotho beta Protein Human (HEK293 His) is the recombinant human-derived Klotho beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-95341 Supplier Medchemexpress LLC Supplier No. HYP7797210UG
Klotho beta protein plays a critical role in the transcriptional repression of cholesterol 7-alpha-hydroxylase (CYP7A1) a key enzyme controlling bile acid synthesis Although its glycosidase activity may be inactive Klotho beta enhances the binding ability of FGFR1 and FGFR4 to FGF21 promoting their interaction Klotho beta Protein Human (HEK293 His) is the recombinant human-derived Klotho beta protein expressed by HEK293 with C-His labeled tag