Learn More
Aviva Systems Biology Corp KLRC1 Antibody - N-terminal region (ARP61289_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61289_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLRC1. It was validated on Western Blot. . Alternate names: CD159A, MGC13374, MGC59791, NKG2, NKG2AApplicationWB. Species reactivity: Guinea Pig, Human, Mouse
Catalog No. 50-287-331