Learn More
Aviva Systems Biology Corp KLRC3 Antibody - N-terminal region (ARP45260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLRC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KLRC3. Alternate names: NKG2-E, NKG2EApplicationWB. Species reactivity: Human, Mouse
Catalog No. 50-014-58992