Learn More
Aviva Systems Biology Corp KLRF1 Antibody - middle region (ARP47263_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47263_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KLRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KLRF1. Alternate names: CLEC5C, MGC119907, MGC119908, MGC119909, NKp80ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59502