KLRG1 is a recombinant human extracellular domain provided as a lyophilized protein with N-terminal SUMO and 6xHis tags. It is expressed in E. coli, purified to >95% by reducing SDS-PAGE, and formulated in Tris-HCl/EDTA/trehalose for stability. The product corresponds to residues L60-F195 (Accession Q96E93) and is intended for biochemical and cell-based studies of KLRG1 binding and inhibitory signaling.
- Recombinant human extracellular domain with N-terminal SUMO and 6xHis tag.
- Expressed in E. coli for efficient production.
- Purified to greater than 95% by reducing SDS-PAGE.
- Lyophilized formulation with trehalose for enhanced stability.
- Contains residues L60-F195 (Accession Q96E93) for native binding activity.
- Suitable for biochemical and cell-based assays of receptor-ligand interactions.
- Stable when stored at -20°C; aliquot and freeze for long-term storage.