Learn More
Medchemexpress LLC KLRG1 Human His-SU 50ug
SDP
Catalog No. 5000196714
Encompass_Preferred
Recombinant human KLRG1 extracellular fragment expressed in E. coli with N-terminal SUMO and 6xHis tags. Supplied as a lyophilized powder formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), the product is provided at 50 μg, with >95% purity and endotoxin <1 EU/μg. Intended for immunology and cell signaling research involving NK and T cell inhibition and cadherin interactions.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for many cell-based assays.
  • N-SUMO and N-6xHis tags facilitate purification and detection.
  • Lyophilized formulation with trehalose for enhanced stability during storage.
  • Contains human extracellular sequence L60-F195, relevant for cadherin binding studies.
  • Recommended reconstitution concentration: not less than 100 μg/mL in ddH2O.
Catalog No. 50-001-96714 Supplier Medchemexpress LLC Supplier No. HYP7160750UG
Recombinant human KLRG1 extracellular fragment expressed in E. coli with N-terminal SUMO and 6xHis tags. Supplied as a lyophilized powder formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), the product is provided at 50 μg, with >95% purity and endotoxin <1 EU/μg. Intended for immunology and cell signaling research involving NK and T cell inhibition and cadherin interactions.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for many cell-based assays.
  • N-SUMO and N-6xHis tags facilitate purification and detection.
  • Lyophilized formulation with trehalose for enhanced stability during storage.
  • Contains human extracellular sequence L60-F195, relevant for cadherin binding studies.
  • Recommended reconstitution concentration: not less than 100 μg/mL in ddH2O.