Learn More
Cayman Chemical KM 233 Neurochemicals
SDP
Catalog No. 502890690
Encompass_Preferred
Δ8-THC analog with 13-fold selectivity for the CB2 receptor (Kis = 12.3 and 0.91 nM, for CB1 and CB2 respectively); its CB agonist activity inhibits human U87 glioma cell proliferation in vitro ( IC50 = 1.4 μM) and in vivo (2 mg/kg in a SCID mouse xenograft side-pocket model)
Catalog No. 50-289-0690 Supplier Cayman Chemical Supplier No. 106401 MG
Δ8-THC analog with 13-fold selectivity for the CB2 receptor (Kis = 12.3 and 0.91 nM, for CB1 and CB2 respectively); its CB agonist activity inhibits human U87 glioma cell proliferation in vitro ( IC50 = 1.4 μM) and in vivo (2 mg/kg in a SCID mouse xenograft side-pocket model)