Learn More
Medchemexpress LLC HY-19970 10mg Medchemexpress, KM11060 CAS:774549-97-2 Purity:>98%
SDP
Catalog No. 502082203
Encompass_Preferred
Medchemexpress, HY-19970 10mg KM11060 CAS:774549-97-2 KM11060 is a corrector of the F508 deletion (F508del)-cystic fibrosis transmembrane conductance regulator (CFTR) trafficking defect. KM11060 can be used for the research of F508del-CFTR processing defect and development of cystic fibrosis therapeutics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2203 Supplier Medchemexpress LLC Supplier No. HY1997010MG
Medchemexpress, HY-19970 10mg KM11060 CAS:774549-97-2 KM11060 is a corrector of the F508 deletion (F508del)-cystic fibrosis transmembrane conductance regulator (CFTR) trafficking defect. KM11060 can be used for the research of F508del-CFTR processing defect and development of cystic fibrosis therapeutics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.