Learn More
Medchemexpress LLC HY-120139 5mg Medchemexpress, KMH-233 CAS:1941174-13-5 Purity:>98%
SDP
Catalog No. 502080935
Encompass_Preferred
Medchemexpress, HY-120139 5mg KMH-233 CAS:1941174-13-5 KMH-233, a potent, reversible and selective l-type amino acid transporter 1 (LAT1) inhibitor, inhibits the uptake of LAT1 substrate, l-leucin (IC50=18 μM) as well as cell growth. KMH-233 significantly potentiates the efficacy of Bestatin and Cisplatin even at low concentrations (25 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0935 Supplier Medchemexpress LLC Supplier No. HY1201395MG
Medchemexpress, HY-120139 5mg KMH-233 CAS:1941174-13-5 KMH-233, a potent, reversible and selective l-type amino acid transporter 1 (LAT1) inhibitor, inhibits the uptake of LAT1 substrate, l-leucin (IC50=18 μM) as well as cell growth. KMH-233 significantly potentiates the efficacy of Bestatin and Cisplatin even at low concentrations (25 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.