Learn More
Aviva Systems Biology Corp KMO Antibody - N-terminal region (ARP64018_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64018_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KMO. It was validated on Western Blot. . Alternate names: dJ317G22.1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-143