Learn More
Enzo Life Sciences KN-62 (1mg). CAS: 127191-97-3
SDP
Supplier:  Enzo Life Sciences BMLEI2300001
Encompass_Preferred
KN-62 is a potent and selective inhibitor of Ca2+/calmodulin kinase II (Ki=0.9 µM). Causes cells to arrest in S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 µM blocked CaMKII phosphorylation of III-AC and inhibited hormone-stimulated Ca2+ oscillations. Purity: ≥98% (TLC). Solubility: Soluble in methanol and DMSO (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0623