Learn More
Apexbio Technology LLC KN-92 hydrochloride 1431698-47-3 200mg
SDP
Catalog No. 5000093746
Encompass_Preferred
KN-92 hydrochloride is an inactive structural analog of the Ca /calmodulin-dependent protein kinase II (CaMKII) inhibitor KN-93 serving primarily as a comparative control compound in biological research Unlike KN-93 KN-92 hydrochloride lacks meaningful inhibitory activity against CaMKII exhibiting a much higher Ki value ( 100 M) In experimental models it is utilized to distinguish CaMKII-dependent molecular and physiological responses particularly in studies of early afterdepolarizations (EADs) and cardiac repolarization events Based on these properties KN-92 hydrochloride is applied as a negative control in cardiac electrophysiological research
Catalog No. 50-000-93746 Supplier Apexbio Technology LLC Supplier No. A3530200
KN-92 hydrochloride is an inactive structural analog of the Ca /calmodulin-dependent protein kinase II (CaMKII) inhibitor KN-93 serving primarily as a comparative control compound in biological research Unlike KN-93 KN-92 hydrochloride lacks meaningful inhibitory activity against CaMKII exhibiting a much higher Ki value ( 100 M) In experimental models it is utilized to distinguish CaMKII-dependent molecular and physiological responses particularly in studies of early afterdepolarizations (EADs) and cardiac repolarization events Based on these properties KN-92 hydrochloride is applied as a negative control in cardiac electrophysiological research