Learn More
Apexbio Technology LLC KN-92 phosphate 1135280-28-2 5mg
SDP
Catalog No. 5000093625
Encompass_Preferred
KN-92 phosphate is a small-molecule analog of the Ca2 /calmodulin-dependent kinase II (CaMKII) inhibitor KN-93 It is designed as an inactive control compound to distinguish CaMKII-specific effects in experimental settings KN-92 phosphate does not inhibit CaMKII activity and thus serves as a negative control for CaMKII inhibition studies Additionally KN-92 phosphate blocks certain voltage-gated potassium (Kv) channels such as Kv1 2 and Kv1 5 reducing steady-state current amplitudes upon depolarization Based on these pharmacological properties KN-92 phosphate holds research potential in investigating mechanisms underlying CaMKII-related signaling pathways and Kv channel functions
Catalog No. 50-000-93625 Supplier Apexbio Technology LLC Supplier No. A35315
KN-92 phosphate is a small-molecule analog of the Ca2 /calmodulin-dependent kinase II (CaMKII) inhibitor KN-93 It is designed as an inactive control compound to distinguish CaMKII-specific effects in experimental settings KN-92 phosphate does not inhibit CaMKII activity and thus serves as a negative control for CaMKII inhibition studies Additionally KN-92 phosphate blocks certain voltage-gated potassium (Kv) channels such as Kv1 2 and Kv1 5 reducing steady-state current amplitudes upon depolarization Based on these pharmacological properties KN-92 phosphate holds research potential in investigating mechanisms underlying CaMKII-related signaling pathways and Kv channel functions