Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KN-92 phosphate | 1135280-28-2 | MFCD24369012 | 99.7% | 554.98 | C24H28ClN2O7PS | 10 MG
SDP

Catalog No. 5000287359
Encompass_Preferred

KN-92 phosphate is the inactive phosphate derivative of a CaMKII inhibitor, provided as a white to off-white solid for use as a control compound in biochemical and cellular studies. It is characterized by high purity and defined physicochemical properties, with limited water solubility and recommended sealed storage away from moisture.

  • Intended as a negative control for studies of CaMKII activity.
  • High purity (~99.7%) suitable for research applications.
  • Molecular weight 554.98; formula C24H28ClN2O7PS.
  • Low solubility in water; soluble in DMSO (50 mg/mL).
  • Packaged as a 10 mg quantity for laboratory use.
  • For research use only; not intended for human use.

Catalog No. 50-002-87359 Supplier Medchemexpress LLC Supplier No. HY15517A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KN-92 phosphate is the inactive phosphate derivative of a CaMKII inhibitor, provided as a white to off-white solid for use as a control compound in biochemical and cellular studies. It is characterized by high purity and defined physicochemical properties, with limited water solubility and recommended sealed storage away from moisture.

  • Intended as a negative control for studies of CaMKII activity.
  • High purity (~99.7%) suitable for research applications.
  • Molecular weight 554.98; formula C24H28ClN2O7PS.
  • Low solubility in water; soluble in DMSO (50 mg/mL).
  • Packaged as a 10 mg quantity for laboratory use.
  • For research use only; not intended for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.