Learn More
Selleck Chemical LLC KN-92 phosphate
SDP
Catalog No. 5000002988
Encompass_Preferred
KN-92 is an inactive derivative of KN-93 the selective inhibitor of Ca2 /calmodulin-dependent kinase type II (CaMKII)
Catalog No. 50-000-02988 Supplier Selleck Chemical LLC Supplier No. S65072MG
KN-92 is an inactive derivative of KN-93 the selective inhibitor of Ca2 /calmodulin-dependent kinase type II (CaMKII)