Learn More
Selleck Chemical LLC KN-92 phosphate
SDP
Supplier:  Selleck Chemical LLC S65075MG
Encompass_Preferred
KN-92 is an inactive derivative of KN-93 the selective inhibitor of Ca2 /calmodulin-dependent kinase type II (CaMKII)
Catalog No. 50-000-03454