Learn More
TARGETMOL CHEMICALS INC KN-92 PHOSPHATE 5MG
SDP
Catalog No. 502576754
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KN-92 phosphate (KN92-H3PO4) is an inactive derivative of KN-93. KN-93 is a selective inhibitor of Ca2+/calmodulin-dependent kinase II (CaMKII) competitively blocking CaM binding to the kinase (Ki = 370 nM). purity: 99%
Catalog No. 50-257-6754 Supplier TARGETMOL CHEMICALS INC Supplier No. T45305MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KN-92 phosphate (KN92-H3PO4) is an inactive derivative of KN-93. KN-93 is a selective inhibitor of Ca2+/calmodulin-dependent kinase II (CaMKII) competitively blocking CaM binding to the kinase (Ki = 370 nM). purity: 99%