Learn More
Cayman Chemical KN-92hydrochlorIde 10mg
SDP
Catalog No. 502892451
Encompass_Preferred
An inactive derivative of KN-93, the selective inhibitor of CaMKII; ineffective at inhibiting CaMKII or arresting cell growth of NIH 3T3 fibroblasts at concentrations up to 25 μM
Catalog No. 50-289-2451 Supplier Cayman Chemical Supplier No. 900089010 MG
An inactive derivative of KN-93, the selective inhibitor of CaMKII; ineffective at inhibiting CaMKII or arresting cell growth of NIH 3T3 fibroblasts at concentrations up to 25 μM