Learn More
Medchemexpress LLC KN-93 | 139298-40-1 | C26H29ClN2O4S | 25 MG
SDP
Catalog No. 5000375209
Encompass_Preferred
KN-93 is a cell-permeable, reversible, and competitive inhibitor of calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. It is for research use only.
  • Cell-permeable, reversible, and competitive inhibitor of CaMKII
  • Ki of 370 nM
  • High purity of 99.42%
  • Appears as a white to off-white solid
  • Blocks cell growth stimulated by growth factors
  • Inhibits H+, K+-ATPase activity
  • Inhibits activation of CaMKII/NF-κB signaling
  • Inhibits retinal vascular leakage in diabetic models
Catalog No. 50-003-75209 Supplier Medchemexpress LLC Supplier No. 25MGHY15465
KN-93 is a cell-permeable, reversible, and competitive inhibitor of calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. It is for research use only.
  • Cell-permeable, reversible, and competitive inhibitor of CaMKII
  • Ki of 370 nM
  • High purity of 99.42%
  • Appears as a white to off-white solid
  • Blocks cell growth stimulated by growth factors
  • Inhibits H+, K+-ATPase activity
  • Inhibits activation of CaMKII/NF-κB signaling
  • Inhibits retinal vascular leakage in diabetic models