Learn More
Enzo Life Sciences KN-93 (1mg). CAS: 139298-40-1
SDP
Supplier:  Enzo Life Sciences BMLEI2680001
Encompass_Preferred
KN-93 is a potent, selective and cell-permeable inhibitor of CaM kinase II (IC50=370 nM). Does not significantly inhibit PKA. Causes cell-cycle arrest in NIH 3T3 fibroblasts and HeLa cells. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0649