Learn More
Medchemexpress LLC HY-15465 1mg , KN-93 CAS:139298-40-1 Purity:>98%
SDP
Catalog No. 502260164
Encompass_Preferred
HY-15465 1mg KN-93 CAS: 139298-40-1 KN-93 is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0164 Supplier Medchemexpress LLC Supplier No. HY154651MG
HY-15465 1mg KN-93 CAS: 139298-40-1 KN-93 is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.