Learn More
Apexbio Technology LLC KN-93 139298-40-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063002
Encompass_Preferred
KN-93 (CAS 139298-40-1) is a selective inhibitor of calcium/calmodulin-dependent protein kinase II (CaMKII) a serine/threonine kinase activated by calcium-bound calmodulin KN-93 inhibits CaMKII enzymatic activity with reported IC50 values of approximately 0 37 0 5 M in vitro and a Ki value of 2 58 M against cardiac CaMKII In NIH 3T3 cells KN-93 (12 M) induces G1 cell-cycle arrest and at higher concentrations (24 M) promotes apoptosis Due to CaMKII s role in neuronal function cardiac signaling immune activation and disease states including Alzheimer s and arrhythmias KN-93 serves as a tool compound for studying CaMKII-mediated processes in biomedical research
Catalog No. 50-000-63002 Supplier Apexbio Technology LLC Supplier No. A35325.1
KN-93 (CAS 139298-40-1) is a selective inhibitor of calcium/calmodulin-dependent protein kinase II (CaMKII) a serine/threonine kinase activated by calcium-bound calmodulin KN-93 inhibits CaMKII enzymatic activity with reported IC50 values of approximately 0 37 0 5 M in vitro and a Ki value of 2 58 M against cardiac CaMKII In NIH 3T3 cells KN-93 (12 M) induces G1 cell-cycle arrest and at higher concentrations (24 M) promotes apoptosis Due to CaMKII s role in neuronal function cardiac signaling immune activation and disease states including Alzheimer s and arrhythmias KN-93 serves as a tool compound for studying CaMKII-mediated processes in biomedical research