Learn More
Selleck Chemical LLC KN-93
SDP
Catalog No. 5000002501
Encompass_Preferred
KN-93 is a cell-permeable reversible and competitive inhibitor of Ca2 /calmodulin-dependent protein kinase II (CaMK-II) with Ki of 2 58 M at rabbit myocardial CaM kinase activity
Catalog No. 50-000-02501 Supplier Selleck Chemical LLC Supplier No. S678725MG
KN-93 is a cell-permeable reversible and competitive inhibitor of Ca2 /calmodulin-dependent protein kinase II (CaMK-II) with Ki of 2 58 M at rabbit myocardial CaM kinase activity