Learn More
Medchemexpress LLC KN-93 hydrochloride | 1956426-56-4 | C26H30Cl2N2O4S | 25 MG
SDP
Catalog No. 5000375210
Encompass_Preferred
KN-93 hydrochloride is a cell-permeable, reversible, and competitive inhibitor of calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. It is intended for research use only.
  • Cell-permeable, reversible, and competitive CaMKII inhibitor
  • Ki of 370 nM
  • Arrests cells in G1 phase
  • Blocks cell growth stimulated by basic fibroblast growth factor, platelet-derived growth factor-BB, and epidermal growth factor
  • Inhibits H+, K+-ATPase activity
  • Prevents increased LV developed pressure during action potential prolongation
Catalog No. 50-003-75210 Supplier Medchemexpress LLC Supplier No. 25MGHY15465A
KN-93 hydrochloride is a cell-permeable, reversible, and competitive inhibitor of calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. It is intended for research use only.
  • Cell-permeable, reversible, and competitive CaMKII inhibitor
  • Ki of 370 nM
  • Arrests cells in G1 phase
  • Blocks cell growth stimulated by basic fibroblast growth factor, platelet-derived growth factor-BB, and epidermal growth factor
  • Inhibits H+, K+-ATPase activity
  • Prevents increased LV developed pressure during action potential prolongation