Learn More
Medchemexpress LLC HY-15465A 5mg Medchemexpress, KN-93 (hydrochloride) CAS:1956426-56-4 Purity:>98%
SDP
Catalog No. 502030633
Encompass_Preferred
Medchemexpress, HY-15465A 5mg KN-93 (hydrochloride) CAS:1956426-56-4 KN-93 hydrochloride is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0633 Supplier Medchemexpress LLC Supplier No. HY15465A5MG
Medchemexpress, HY-15465A 5mg KN-93 (hydrochloride) CAS:1956426-56-4 KN-93 hydrochloride is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.