Learn More
TARGETMOL CHEMICALS INC KN-93 PHOSPHATE 10MG
SDP
Catalog No. 502580723
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II with K i of 370 nM. KN-93 phosphate inhibits serum-induced fibroblast cell growth in a comparable dose-dependent fashion to its inhibition of CaMK-II activity. purity: 99%
Catalog No. 50-258-0723 Supplier TARGETMOL CHEMICALS INC Supplier No. T260610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II with K i of 370 nM. KN-93 phosphate inhibits serum-induced fibroblast cell growth in a comparable dose-dependent fashion to its inhibition of CaMK-II activity. purity: 99%