Learn More
Medchemexpress LLC Kn-93 phosphate | 1913269-12-1 | 99.7% | 599.03 g·mol⁻¹ | C26H32ClN2O8PS | 10 MG
SDP
Catalog No. 5000286631
Encompass_Preferred
KN-93 phosphate is the phosphate salt form of KN-93, a cell-permeable, reversible, competitive inhibitor of calmodulin-dependent kinase II (CaMKII) with a reported Ki of 370 nM. It is provided as a high-purity research reagent intended for in vitro and cell-based studies investigating CaMKII signaling pathways.
  • High purity suitable for research applications.
  • Water-soluble phosphate salt for improved solubility in aqueous assays.
  • Reported Ki of 370 nM against CaMKII for potency reference.
  • Available in small pack sizes to support screening and optimization.
  • Provided with analytical data supporting quality and identity.
Catalog No. 50-002-86631 Supplier Medchemexpress LLC Supplier No. HY15465B10MG
KN-93 phosphate is the phosphate salt form of KN-93, a cell-permeable, reversible, competitive inhibitor of calmodulin-dependent kinase II (CaMKII) with a reported Ki of 370 nM. It is provided as a high-purity research reagent intended for in vitro and cell-based studies investigating CaMKII signaling pathways.
  • High purity suitable for research applications.
  • Water-soluble phosphate salt for improved solubility in aqueous assays.
  • Reported Ki of 370 nM against CaMKII for potency reference.
  • Available in small pack sizes to support screening and optimization.
  • Provided with analytical data supporting quality and identity.