Learn More
Enzo Life Sciences KN-93 phosphate (1mg). CAS: 1188890-41-6
SDP
Supplier:  Enzo Life Sciences ALX430127M001
Encompass_Preferred
Selective inhibitor of rat brain Ca2+/calmodulin-dependent protein kinase II. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A (PKA) activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells. Alternative name: KN-93 (water soluble), 2-[N-(2-Hydroxyethyl)-N-(4-methoxybenzenesulfonyl)]amino-N-(4-chlorocinnamyl)-N-methylbenzylamine . hydrogen phosphate. Purity: ≥98% (1H-NMR). Appearance: White to off-white solid. Solubility: Soluble in DMSO or water. Long Term Storage: -20°C.
Catalog No. 50-200-9214