Learn More
Medchemexpress LLC HY-15465B 1mg Medchemexpress, KN-93 (phosphate) CAS:1913269-12-1 Purity:>98%
SDP
Catalog No. 501932742
Encompass_Preferred
Medchemexpress, HY-15465B 1mg KN-93 (phosphate) CAS:1913269-12-1 KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II , with K i of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2742 Supplier Medchemexpress LLC Supplier No. HY15465B1MG
Medchemexpress, HY-15465B 1mg KN-93 (phosphate) CAS:1913269-12-1 KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II , with K i of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.