Learn More
Selleck Chemical LLC KN-93 Phosphate
SDP
Supplier:  Selleck Chemical LLC S74231G
Encompass_Preferred
KN-93 Phosphate is a potent and specific inhibitor of Ca2 /calmodulin-dependent protein kinase II (CaMKII) with Ki of 0 37 M no remarkable inhibitory effects on APK PKC MLCK or Ca2 -PDE activities KN-93 attenuates CaMKII-induced autophagy This product is not soluble in saline Please do not dissolve it in saline for administration
Catalog No. 50-000-03538