Learn More
AdipoGen KN-93 (water soluble) (1 mg)
SDP
Catalog No. 501490063
Encompass_Preferred
KN-93 (water soluble) (1 mg), AdipoGen Life Sciences. Chemical. CAS: 1188890-41-6 | 139298-40-1 (free base). Formula: C26H29ClN2O4S . H3PO4. MW: 501.0 . 98.0. Selective Ca2+/calmodulin-dependent protein kinase II inhibitor. Insulin release inhibitor. Acid secretion inhibitor. G1 cell cycle arrest inducer. Apoptosis inducer. Ion channel inhibitor. ROS inducer. Androgen receptor inhibitor.
Catalog No. 50-149-0063 Supplier AdipoGen Supplier No. AGCR10065M001
KN-93 (water soluble) (1 mg), AdipoGen Life Sciences. Chemical. CAS: 1188890-41-6 | 139298-40-1 (free base). Formula: C26H29ClN2O4S . H3PO4. MW: 501.0 . 98.0. Selective Ca2+/calmodulin-dependent protein kinase II inhibitor. Insulin release inhibitor. Acid secretion inhibitor. G1 cell cycle arrest inducer. Apoptosis inducer. Ion channel inhibitor. ROS inducer. Androgen receptor inhibitor.