Learn More
Cayman Chemical KN-93hydrochlorIde 10mg
SDP
Catalog No. 502955745
Encompass_Preferred
A selective inhibitor of CaMKII, competitively blocking CaM binding to the kinase (Ki = 370 nM); inhibits histamine-induced aminopyrine uptake in parietal cells (IC50 = 300 nM)
Catalog No. 50-295-5745 Supplier Cayman Chemical Supplier No. 1386410 MG
A selective inhibitor of CaMKII, competitively blocking CaM binding to the kinase (Ki = 370 nM); inhibits histamine-induced aminopyrine uptake in parietal cells (IC50 = 300 nM)