Learn More
Dynamic Diagnostics KN95 Face Mask With Comfort Earloops
Catalog No. 22899687
Change view
Click to view available options
Non-sterile, single-use KN95 face masks; Individually wrapped
Catalog No.
22-899-687 Non-sterile, single-use KN95 face masks; Individually wrapped
1 options
Catalog No. 22-899-687 Supplier Dynamic Diagnostics Supplier No. 10147
Non-sterile, single-use KN95 face masks. Individually wrapped.
  • Adjustable nose band with cushion
  • Comfortable ear loops
  • Close fit to minimize air gaps
  • 4 layers of protection
  • Particle Filtration Efficiency: > 95%
  • Bacterial Filtration Efficiency: > 95%
Specifications
Color White
Disposable Single-use
Product Type General Purpose Particulate Respirator
Size Standard
Filter Efficiency 95%
Latex Free Yes
Model 10147
Sterility Non-sterile